missing translation for 'onlineSavingsMsg'
Learn More

3M™ Disposable Protective Coveralls with Collar, 4510 Series

Catalog No. 19164136
Change view
Click to view available options
Size:
Large
X-Large
3X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19164136 3X-Large
19164138 Large
17624101 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19164136 Supplier 3M™ Supplier No. 7000088922
Only null left

Coveralls provide protection from certain light liquid splashes and hazardous dusts while enhancing movement and comfort.

  • Elastic waist, ankles, and wrists for convenience and freedom of movement
  • Two-way zipper with storm flap for convenience and additional protection
  • Anti-static coating on both sides helps reduce static build-up and prevent the risk of static discharge
  • Does not contain components made from natural rubber latex to help prevent reactions from users with latex sensitivities or restrictions
  • Does not contain components made from silicone to help prevent reactions from users with sensitivities or restrictions

Specifications

Product Type General Purpose Coveralls
Size 3X-Large
Gender Unisex
Material Laminate
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper with Storm Flap
Seam Type Serged
Sterility Non-sterile
Resistance Anti-static, Dust, Liquid
Certifications/Compliance (Europe) CE, EN 1149-1:2006, EN ISO 13982-1 Type 5, EN 13034 Type 6
Latex Free Yes
Model 4510CS-BLK
Size (English) Chest 49 to 52 in.
Size (English) Inseam 35 in.
Size (English) Waist 54 in.
Size (Metric) Chest 124.46 to 132.08 cm
Size (Metric) Inseam 88.9 cm
Size (Metric) Waist 137.16 cm
For Use With (Application) Abatement, Remediation, Cleaning, Maintenance, Food Processing, Painting, Pesticide, Pharmaceutical
Height (English) 6 to 6.33 ft.
Height (Metric) 186 to 194 cm
Show More Show Less