missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tians Disposable Polypropylene Coveralls
Low lint coverall for use in ISO cleanroom class 7 critical environments and many industrial applications. Nonwoven fabric is cloth-like and cool to wear.
$131.64 - $145.36
Specifications
| Product Type | Cleanroom Coveralls |
|---|---|
| Material | Polypropylene |
| Color | White |
| Disposable | Single-use |
| Ankle Style | Elastic |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
19-130-2073D
|
Tians
210882XL |
X-Large |
Case of 25 Each for $131.64
|
|
|||||
|
19-130-2073E
|
Tians
2108822X |
2X-Large |
Case of 25 Each for $145.36
|
|
|||||
|
19-130-2073H
|
Tians
2108825X |
5X-Large |
Case of 25 Each for $136.97
|
|
|||||
Description
- Economical white polypropylene coverall is ideal low-cost alternative for general protection in nonhazardous environments
- Cool, comfortable, and lightweight
- Zipper front, elastic wrist and ankles, and folded collar
Specifications
| Cleanroom Coveralls | |
| White | |
| Elastic | |
| Collared | |
| Non-sterile | |
| Class 10,000 (ISO 7) |
| Polypropylene | |
| Single-use | |
| Elastic | |
| Front Zipper | |
| Controlled Environments |