missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Disposable Microscope Slides
Durable plastic microscope slides.
Supplier: Fisherbrand™ 20305220
Description
Durable plastic microscope slides.
- Precleaned and packed per dozen
- Size: 3 × 1 in. (7.6 × 2.5cm)
Specifications
| Plastic | |
| 75 mm | |
| 144/Pk. |
| Yes | |
| 25 mm |