missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Disposable IR Cards, 5x10cm, KBr window, 9.5mm aperture

Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.

$194.99 - $435.23

Specifications

Description Disposable IR Cards
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA43937KJ
View Documents Promotion Details
Thermo Scientific Chemicals
043937KJ
5 Pk.
Each for $194.99
Only null left
 
AA43937KM
View Documents Promotion Details
Thermo Scientific Chemicals
043937KM
25 Pk.
Each for $435.23
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Size: 5 x 10cm
  • KBr window
  • 9.5mm aperture
Specifications

Specifications

Disposable IR Cards
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only