Learn More
GVS Disposable Inoculating Needle
Description
- Smooth Surface – Ensures even and consistent streaking
- Color-Coded Sizes – Allows quick and easy identification
- Ergonomic Polygonal Shaft – Provides a secure grip for easy operation and precise directional control
- Loop Options – Available in both rigid and flexible types upon request
- Secure Packaging – Supplied in individual peel packs (easy peel-open) and zip-lock packs (tamperproof and re-sealable) to minimize the risk of contamination
Specifications
Specifications
| Material | Polypropylene |
| Sterility | Sterile |
| Size | Standard |
| Color | Violet |
| Product Type | Inoculating Needle |
| Packaging Type | 10/Zip-Lock Pack |