Learn More
Fisherbrand™ Disposable Inoculating Loops
Catalog No. 01189168
Change view
Click to view available options
Volume:
10 μL
Sterility:
Sterile
Quantity:
Cs of 1000 (40 Pks. of 25)
Catalog No. Volume Sterility Quantity
01189168 10 μL Sterile Cs of 1000 (40 Pks. of 25)
1 options
Catalog No. 01189168 Supplier Fisherbrand™ Supplier No. 10401285
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These Disposable Inoculating Loops are smooth and flexible to facilitate smooth, uniform streaking without damaging gel surfaces. Sterile and non-sterile options available.
  • Ideal for anaerobic microbiology applications where deep agar inoculation is required
  • Use in situations where flaming is not practical, such as under hoods and in anaerobic chamber
  • High-impact polystyrene loops are smooth and flexible to facilitate smooth, uniform streaking without damaging the gel surface
  • Rigid enough to penetrate the media for subsurface inoculation
  • Disposable loops do not need flaming, thus help to eliminate the risk of infection due to aerosol formation of pathogenic substances
  • Can be used under hoods without danger
  • Color-coded for ease of size identification
Specifications
Disposable Single-use
Material Polystyrene
Volume 10 μL
Sterility Sterile
Quantity Cs of 1000 (40 Pks. of 25)
Autoclavable Not Autoclavable