Learn More
Innovatek Disposable Haemocytometer, Neubauer Improved
Catalog No. 29801080
Change view
Click to view available options
Product Type:
Hemacytometer
Catalog No. Product Type
29801080 Hemacytometer
1 options
Catalog No. 29801080 Supplier Innovatek Supplier No. 4230DHCNO110
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Innovatek Disposable Haemocytometers greatly reduce the risk of exposure to biohazardous fluids
  • Precision, repeatability and reliability
  • Greatly reduces risk of exposure to biohazardous fluid
  • Lightweight, unbreakable
  • Two separate counting chambers on each slide
  • Integrated coverslip of optically clear plastic
Specifications
Product Type Hemacytometer
Disposable Single-use
Material Plastic
Quantity 10 Slides/Box