Learn More
Research Products Corporation Disposable Universal Electroporation Cuvettes, 2 mm Gap, 400 µl Capacity, Blue Round Lid, 50 per Case
Catalog No. 501537655
Encompass
Disposable Universal Electroporation Cuvettes are designed for use with bacterial, yeast, fungi, and mammalian cells. Cuvettes are manufactured from the clearest, highest quality, durable, polycarbonate material and have demonstrated optimal transfection efficiencies over decades of laboratory use. Compatible with any Electroporators or devices which require the use of electroporation cuvettes (Eporator, BioRad, Gene Pulsar and BTX).
Precision built to give exacting tolerances and ensure reproducible results. Cuvettes are skillfully crafted and polished aluminum plates reduce static and arcing, delivering optimum performance. Individually wrapped and gamma irradiated to guarantee sterility. Individually color-coded cuvette lids identify application, with corresponding gap widths.
Catalog No. 501537655 Supplier Research Products Corporation Supplier No. 212631
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Disposable Universal Electroporation Cuvettes are designed for use with bacterial, yeast, fungi, and mammalian cells. Cuvettes are manufactured from the clearest, highest quality, durable, polycarbonate material and have demonstrated optimal transfection efficiencies over decades of laboratory use. Compatible with any Electroporators or devices which require the use of electroporation cuvettes (Eporator, BioRad, Gene Pulsar and BTX).
Precision built to give exacting tolerances and ensure reproducible results. Cuvettes are skillfully crafted and polished aluminum plates reduce static and arcing, delivering optimum performance. Individually wrapped and gamma irradiated to guarantee sterility. Individually color-coded cuvette lids identify application, with corresponding gap widths.