missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Disposable Cleanroom Socks
GREENER_CHOICE

Catalog No. 128960035
Change view
Click to view available options
Size:
11 to 15
6 to 10
Color:
White, Blue Stitching
White, Red Stitching
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color
128960035 6 to 10 White, Blue Stitching
128960036 11 to 15 White, Red Stitching
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 128960035 Supplier Fisherbrand™ Supplier No. 128960035
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Disposable cleanroom socks are specialized for critical environments and are a durable, cost-effective option for cleanroom and extended wear.

  • Fisherbrand Disposable Cleanroom Socks are a cost-effective and cleanroom compatible option for cleanroom and extended wear.
  • Laundered and packaged in ISO Class 5 Cleanrooms, with extremely low particulate contamination
  • The structure of the socks allows high wear-resistance and durability while maintaining good elasticity.
  • Abrasion-resistant, soft, and comfortable to wear.
  • The product should be stored in a ventilated, dry, mildew-proof and insect-proof place.
  • Avoid prolonged product exposure to sunlight.
  • Do not rub against rough objects directly.
  • Launder using a neutral or slightly alkaline (pH <9) synthetic detergent on a gentle cycle to prevent mechanical and chemical damage
  • If the socks are torn or the yarn begins to falls off or tear in multiple places, replace them.

Specifications

Certifications/Compliance ISO Class 5 (Class 100)
Color White, Blue Stitching
For Use With (Application) Cleanroom and other critical environments
Gender Unisex
Height (Metric) 50 cm
Length (Metric) 26 cm
Material Polyester
Size 6 to 10
Weight (Metric) 25 g/pair
Disposable Single-use
Sterility Non-sterile
Packaging Quantity 500 Pair/Cs.
Show More Show Less