Learn More
Thermo Scientific™ Disposable Calibrated Inoculating Loops
Catalog No. R501420
Change view
Click to view available options
Volume:
No Loop
1 μL
10 μL
Product Type:
Inoculating Loop
Inoculating Needle
Color:
Blue
White
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume Product Type Color
R501420 1 μL Inoculating Loop Blue
R501400 No Loop Inoculating Needle White
R501410 10 μL Inoculating Loop Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R501420 Supplier Thermo Scientific™ Supplier No. R501420
Select, inoculate or transfer microbiological sample volume on media plates with these calibrated inoculating loops.
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Disposable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
Eliminate the need for flaming or sterilization with these sterile, disposable loops.
Specifications
Disposable Single-use
Volume 1 μL
Sterility Sterile
Material Plastic (Handle)
Color Blue
Product Type Inoculating Loop
Quantity 30 bags, 32 units/bag
Calibrated Yes
Handle Type Plastic