Learn More
Thermo Scientific™ Disposable Calibrated Inoculating Loops
Select, inoculate or transfer microbiological sample volume on media plates with these calibrated inoculating loops.
Supplier: Thermo Scientific™ R501410
Description
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Disposable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
Eliminate the need for flaming or sterilization with these sterile, disposable loops.
Chemical Identifiers
| Plastic (Handle) | |
| Single-use | |
| Yes |
| Sterile | |
| 30 bags, 32 units/bag | |
| Plastic |
Specifications
| Single-use | |
| Plastic (Handle) | |
| Yellow | |
| 30 bags, 32 units/bag | |
| Plastic |
| 10 μL | |
| Sterile | |
| Inoculating Loop | |
| Yes |