Learn More
Thermo Scientific™ Disposable Calibrated Inoculating Loops
Catalog No. R501400
Change view
Click to view available options
Volume:
No Loop
1 μL
10 μL
Product Type:
Inoculating Loop
Inoculating Needle
Color:
Blue
White
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume Product Type Color
R501400 No Loop Inoculating Needle White
R501410 10 μL Inoculating Loop Yellow
R501420 1 μL Inoculating Loop Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R501400 Supplier Thermo Scientific™ Supplier No. R501400
Select, inoculate or transfer microbiological sample volume on media plates with these calibrated inoculating loops.
Select, inoculate or transfer microbiological sample on media with Thermo Scientific™ Remel™ Disposable Calibrated Inoculating Loops. The gentle, malleable loops offer precise control when selecting or streaking inoculum, without cutting or tearing agar surfaces. The loops are nontoxic, lightweight and are color-coded for easy-recognition and streamlined specimen handling.
Eliminate the need for flaming or sterilization with these sterile, disposable loops.
Specifications
Disposable Single-use
Volume No Loop
Sterility Sterile
Material Plastic (Handle)
Color White
Product Type Inoculating Needle
Quantity 30 bags, 32 units/bag
Calibrated Yes
Handle Type Plastic