Learn More
Epredia™ Disposable Base Molds
Catalog No. 361014337
Change view
Click to view available options
Dimensions (L x W):
15 x 15 mm
24 x 24 mm
30 x 24 mm
37 x 24 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
361014337 24 x 24 mm
361014336 15 x 15 mm
361014338 30 x 24 mm
361014339 37 x 24 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 361014337 Supplier Epredia™ Supplier No. 58951
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use Epredia™ Disposable Base Molds with all standard-size cassettes.
  • Cleaning associated with metal molds is not required
  • Transparency facilitates tissue orientation
Ordering Information:
  • Sold in cases of four boxes.
Specifications
Product Type Base Mold
Material Plastic
Dimensions (L x W) 24 x 24 mm
Disposable Single-use