missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Disposable Aseptic Transfer Cap for Microcarriers Bottles, Sterile
GREENER_CHOICE

Catalog No. 13100506
Change view
Click to view available options
For Use With (Application):
MLL with tubing (1/8 in. ID, 1/4 in. OD)
MPC with tubing (1/4 in. ID, 3/8 in. OD)
For Use With (Equipment):
100 g Microcarrier Bottle
500 g Microcarrier Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) For Use With (Equipment)
13100506 MLL with tubing (1/8 in. ID, 1/4 in. OD) 100 g Microcarrier Bottle
13100507 MPC with tubing (1/4 in. ID, 3/8 in. OD) 100 g Microcarrier Bottle
13100508 MLL with tubing (1/8 in. ID, 1/4 in. OD) 500 g Microcarrier Bottle
13100509 MPC with tubing (1/4 in. ID, 3/8 in. OD) 500 g Microcarrier Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13100506 Supplier Corning™ Supplier No. 4626

Corning microcarriers, available in 100g and 500g bottles, can be fitted with these Corning aseptic transfer caps to prevent adventitious contamination during bioreactor loading.

These are aseptic transfer casp for 100 g and 500 g sizes.

Specifications

Description Disposable 45 mm Aseptic Transfer Cap
For Use With (Application) MLL with tubing (1/8 in. ID, 1/4 in. OD)
For Use With (Equipment) 100 g Microcarrier Bottle
Product Type Aseptic Transfer Screw Cap
Sterility Sterile
Material Polypropylene cap with chemically resistant, heat sealable flexible tubing
Quantity 1/Cs.