Learn More
BrandTech™ BRAND™ Dispensette™ S Organic Fixed-volume Bottletop Dispensers
The Dispensette™ S Organic Fixed Volume Bottletop Dispensers feature a 'floating piston' design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes.
Supplier:  BrandTech™ BRAND™ 4630230
 View more versions of this product
Catalog No. 13-689-054
Only null left
Explore available promotions
Includes: Discharge tube, valve mounting/calibration tool, polypropylene adapters, telescoping filling tube, recirculation tube (optional), performance certificate, operating manual
Description
Description
  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Safe volume setting with internal scalloped track (analog-adjustable instruments)
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replaceable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • Digital and analog-adjustable models
  • DE-M marking
Warranty
One year
Specifications
Specifications
Bottletop Dispenser
90°
ISO 9001
Up to 2.2 g/cm3
1
5 mL
Dispensing organic solvents, including chlorinated and fluorinated hydrocarbons (e.g., trichlorofluoroethane and dichlormethane), acids like concentrated HCl and HNO3 (except HF), trifluoroacetic acid (TFA), tetrahydrofuran (THF) and peroxides
0.1% (5μL)
15°C to 40°C
Up to 500 mm 2/s
0.5% (25μL)
Autoclavable
Gray/Yellow
Without recirculation valve
Borosilicate Glass, Ceramic, ETFE, PFA, PTFE
Analog
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, a mounting tool, and polypropylene adapters (GL 24-25, GL 28/S28, GL 32-33, GL38, S40)
59°F to 104°F
Up to 600 mbar
2 Year
Product Suggestions
Product Suggestions