missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ Dispensette™ S Digital Bottletop Dispensers

The Dispensette™ S Digital Bottletop Dispensers feature a 'floating piston' design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes.
$2,086.91 - $2,981.29
Specifications
| Product Type | Dispenser |
|---|---|
| Angle | 90° |
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certifications: ISO 9001 |
| Color | Gray/Red |
Includes: Discharge tube, valve mounting/calibration tool, polypropylene adapters, telescoping filling tube, recirculation tube (optional), performance certificate, operating manual
| Catalog Number | Mfr. No. | Includes | Volume (Metric) | Increments | Accuracy | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Includes | Volume (Metric) | Increments | Accuracy | Price | Quantity & Availability | ||
13689000
![]() |
BRANDTECH™ BRAND™
4600311 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 24-25, GL 28/S28, GL 32-33, GL38, S40) | 0.1 to 1 mL | 0.005 mL | 0.6% (6μL) |
Each for $2,212.38
|
|
||
13689001
![]() |
BRANDTECH™ BRAND™
4600321 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 24-25, GL 28/S28, GL 32-33, GL38, S40) | 0.2 to 2 mL | 0.01 mL | 0.5% (10μL) |
Each for $2,212.38
|
|
||
13689002
![]() |
BRANDTECH™ BRAND™
4600331 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 24-25, GL 28/S28, GL 32-33, GL38, S40) | 0.5 to 5 mL | 0.02 mL | 0.5% (25μL) |
Each for $2,212.38
|
|
||
13689003
![]() |
BRANDTECH™ BRAND™
4600341 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 24-25, GL 28/S28, GL 32-33, GL38, S40) | 1 to 10 mL | 0.05 mL | 0.5% (50μL) |
Each for $2,212.38
|
|
||
13689004
![]() |
BRANDTECH™ BRAND™
4600351 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40) | 2.5 to 25 mL | 0.1 mL | 0.5% (125μL) |
Each for $2,936.88
|
|
||
13689005
![]() |
BRANDTECH™ BRAND™
4600361 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, recirculation valve with an optional recirculation tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40) | 5 to 50 mL | 0.2 mL | 0.5% (250μL) |
Each for $2,981.29
|
|
||
13689006
![]() |
BRANDTECH™ BRAND™
4600310 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, a mounting tool, and polypropylene adapters (GL 24-25, GL 28/S28, GL 32-33, GL38, S40) | 0.1 to 1 mL | 0.005 mL | 0.6% (6μL) |
Each for $2,086.91
|
|
||
13689007
![]() |
BRANDTECH™ BRAND™
4600320 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, a mounting tool, and polypropylene adapters (GL 24-25, GL 28/S28, GL 32-33, GL38, S40) | 0.2 to 2 mL | 0.01 mL | 0.5% (10μL) |
Each for $2,086.91
|
|
||
13689008
![]() |
BRANDTECH™ BRAND™
4600330 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, a mounting tool, and polypropylene adapters (GL 24-25, GL 28/S28, GL 32-33, GL38, S40) | 0.5 to 5 mL | 0.02 mL | 0.5% (25μL) |
Each for $2,086.91
|
|
||
13689009
![]() |
BRANDTECH™ BRAND™
4600340 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, a mounting tool, and polypropylene adapters (GL 24-25, GL 28/S28, GL 32-33, GL38, S40) | 1 to 10 mL | 0.05 mL | 0.5% (50μL) |
Each for $2,086.91
|
|
||
13689010
![]() |
BRANDTECH™ BRAND™
4600350 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40) | 2.5 to 25 mL | 0.1 mL | 0.5% (125μL) |
Each for $2,809.21
|
|
||
13689011
![]() |
BRANDTECH™ BRAND™
4600360 |
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40) | 5 to 50 mL | 0.2 mL | 0.5% (250μL) |
Each for $2,856.38
|
|
||
Description
The Dispensette™ S Digital Bottletop Dispensers feature a floating piston design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for g
- New discharge tube, with or without recirculation valve
- New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
- Simple priming by optimized flow channels
- Less dispensing force - especially with large volume instruments
- Easy Calibration technique for calibration adjustment in seconds without tools
- Pivoted screw cap - does not disturb dispensing
- Easy to calibrate and adjust in order to comply with GLP guidelines
- Easy to dismantle for cleaning
- Replaceable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
- 360° rotatable valve block allows bottle label to face user
- Telescoping filling tube adjusts easily to different size bottles
- DE-M marking
Warranty
One year
Specifications
| Dispenser | |
| Autoclavable | |
| Gray/Red | |
| Digital | |
| 59°F to 104°F | |
| Up to 500 mm 2/s |
| 90° | |
| Certifications: ISO 9001 | |
| Up to 2.2 g/cm3 | |
| Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents | |
| 15°C to 40°C |