Learn More
BRANDTECH™ BRAND™ Dispensette™ S Analog-adjustable Bottletop Dispensers
GREENER_CHOICE
The Dispensette™ S Analog Bottletop Dispensers feature a 'floating piston' design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes.
Supplier:  BRANDTECH™ BRAND™ 4600160
 View more versions of this product
Catalog No. 13689024
Only null left
Includes: Discharge tube, valve mounting/calibration tool, polypropylene adapters, telescoping filling tube, recirculation tube (optional), performance certificate, operating manual
Description
Description
  • New discharge tube, with or without recirculation valve
  • New valve system - no additional sealing rings required, facilitates cleaning and preparations for autoclaving
  • Simple priming by optimized flow channels
  • Less dispensing force - especially with large volume instruments
  • Safe volume setting with internal scalloped track (analog-adjustable instruments)
  • Pivoted screw cap - does not disturb dispensing
  • Easy to calibrate and adjust in order to comply with GLP guidelines
  • Easy to dismantle for cleaning
  • Replaceable filling and discharge valve with safety bulb (locks when discharge tube is not assembled)
  • 360° rotatable valve block allows bottle label to face user
  • Telescoping filling tube adjusts easily to different size bottles
  • DE-M marking
Warranty
One year
Specifications
Chemical Identifiers
Bottletop Dispenser
Autoclavable
Gray/Red
1
Analog
59°F to 104°F
Up to 600 mbar
2 Year
90°
ISO 9001
Up to 2.2 g/cm3
Borosilicate Glass, Ceramic, ETFE, PFA, PTFE
Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents
15°C to 40°C
Up to 500 mm 2/s
Specifications
0.5% (250μL)
Bottletop Dispenser
ISO 9001
Up to 2.2 g/cm3
1
5 to 50 mL
Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents
1.0 mL
59°F to 104°F
Up to 600 mbar
2 Year
90°
Autoclavable
Gray/Red
Without recirculation valve
Borosilicate Glass, Ceramic, ETFE, PFA, PTFE
Analog
Dispensette S bottle-top dispenser,a GL 45 threaded bottle adapter, DE-M marking, performance certificate, telescoping filling tube, a mounting tool, and polypropylene adapters (GL 32-33, GL38, S40)
0.1% (50 μL)
15°C to 40°C
Up to 500 mm 2/s
Product Suggestions
Product Suggestions