Neutral Protease (Dispase), Its mild proteolytic action makes the enzyme especially suitable for the preparation of primary and secondary (subcultivation) cell culture, A lyophilized powder, Pack Size: 5mg
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Neutral Protease (Dispase), Its mild proteolytic action makes the enzyme especially suitable for the preparation of primary and secondary (subcultivation) cell culture, A lyophilized powder, Pack Size: 5mg