Genuine Tecan 500 L disposable automation tips designed specifically for use on the Freedom EVO and Fluent MultiChannel Arm™ (MCA) 96 option. ANSI format, single stack, filtered. Maximum pipetting volume of 400 L. Tecan STERILE purity level - sterile, tested and certified to be free from human DNA, Dnase, Rnase, pyrogens and endotoxins. 40 tip boxes x 96 tips, 3,840 tips/sales unit. All MultiChannel Arm™(MCA) 96 disposable tips are designed, manufactured, tested and certified to guarantee reliable quality, helping you to acquire optimal results.