Learn More
Paul N Gardner Company DISP CUVETTE 50X10 PLAS W/CAP
Catalog No. NC9781864
Encompass
Rectangular cuvette, 50 x 10mm, PMMA, disposable, 10 pcs for Lico Colorimeter.
Catalog No. NC9781864 Supplier Paul N Gardner Company Supplier No. CL4823
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Rectangular cuvette, 50 x 10mm, PMMA, disposable, 10 pcs for Lico Colorimeter.