missing translation for 'onlineSavingsMsg'
Learn More

Disodium Sebacate 97.0+%, TCI America™

Catalog No. S027025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S027025G 25 g
S0270500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S027025G Supplier TCI America Supplier No. S027025G
Only null left

Chemical Identifiers

CAS 17265-14-4
Molecular Formula C10H16Na2O4
Molecular Weight (g/mol) 246.214
MDL Number MFCD00070501
InChI Key NCXUIEDQTCQZRK-UHFFFAOYSA-L
Synonym Decanedioic Acid Disodium Salt, Sebacic Acid Disodium Salt, Disodium Decanedioate
PubChem CID 134734
IUPAC Name disodium;decanedioate
SMILES C(CCCCC(=O)[O-])CCCC(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 246.21
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Sebacate
RTECSNumber VS1430000
TSCA Yes