missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium Rhodizonate 90.0+%, TCI America™
$80.16 - $807.14
Chemical Identifiers
| CAS | 523-21-7 |
|---|---|
| Molecular Formula | C6Na2O6 |
| Molecular Weight (g/mol) | 214.04 |
| MDL Number | MFCD00001650 |
| InChI Key | SXWPCWBFJXDMAI-UHFFFAOYSA-L |
| Synonym | sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate |
| PubChem CID | 68225 |
| IUPAC Name | disodium tetraoxocyclohex-1-ene-1,2-bis(olate) |
| SMILES | [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O |
Chemical Identifiers
| 523-21-7 | |
| 214.04 | |
| SXWPCWBFJXDMAI-UHFFFAOYSA-L | |
| 68225 | |
| [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O |
| C6Na2O6 | |
| MFCD00001650 | |
| sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate | |
| disodium tetraoxocyclohex-1-ene-1,2-bis(olate) |
Specifications
| 300°C | |
| 523-21-7 | |
| 1 g | |
| MFCD00001650 | |
| SXWPCWBFJXDMAI-UHFFFAOYSA-L | |
| disodium tetraoxocyclohex-1-ene-1,2-bis(olate) | |
| 68225 | |
| ≥90.0% (T) |
| Green-Yellow | |
| Crystalline Powder | |
| C6Na2O6 | |
| sodium rhodizonate, sodium 3,4,5,6-tetraoxocyclohex-1-ene-1,2-bis olate, rhodizonic acid disodium salt, unii-9qf0329l3g, rhodizonic acid sodium salt, rhodizonic acid, disodium salt, disodium 3,4,5,6-tetraoxocyclohexene-1,2-diolate, 3,4,5,6-tetraoxocyclohexene-1,2-diol disodium salt, disodium tetraoxocyclohex-1-ene-1,2-bis olate, dipotassium tetraoxocyclohex-1-ene-1,2-bis olate | |
| [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C(=O)C1=O | |
| 214.04 | |
| 214.04 | |
| Disodium Rhodizonate |
Safety and Handling
TSCA : No