missing translation for 'onlineSavingsMsg'
Learn More

Disodium Phthalocyanine, TCI America™

Catalog No. P09731G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P09731G Supplier TCI America Supplier No. P09731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25476-27-1
Molecular Formula C32H18N8Na2
Molecular Weight (g/mol) 560.532
MDL Number MFCD00066368
InChI Key VISVNSVJALJRHK-UHFFFAOYSA-N
Synonym Phthalocyanine Sodium, Sodium Phthalocyanine, Phthalocyanine Disodium
PubChem CID 66737394
SMILES C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3.[Na].[Na]

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 558.52
Physical Form Crystalline Powder
Chemical Name or Material Disodium Phthalocyanine