missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium Phthalate 95.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 500 g |
| Formula Weight | 210.10 |
| Percent Purity | ≥95.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Disodium Phthalate |
Safety and Handling
| EINECSNumber | (3)-1272 |
| TSCA | Yes |