missing translation for 'onlineSavingsMsg'
Learn More

Disodium Phthalate 95.0+%, TCI America™

Catalog No. P040025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P040025G 25 g
P0400500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P040025G Supplier TCI America Supplier No. P040025G
Only null left

Chemical Identifiers

CAS 15968-01-1
Molecular Formula C8H4Na2O4
Molecular Weight (g/mol) 210.096
MDL Number MFCD00050736
InChI Key HQWKKEIVHQXCPI-UHFFFAOYSA-L
Synonym disodium phthalate, sodium phthalate, phthalic acid disodium salt, unii-qb14yv193c, 1,2-benzenedicarboxylic acid, disodium salt, sodium 2-carboxybenzoate, 1,2-benzenedicarboxylic acid, sodium salt 1:2, 88-99-3 parent, disodiumphthalate, acmc-209dk3
PubChem CID 85215
IUPAC Name disodium;phthalate
SMILES C1=CC=C(C(=C1)C(=O)[O-])C(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 210.10
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Phthalate
EINECSNumber (3)-1272
TSCA Yes