missing translation for 'onlineSavingsMsg'
Learn More

Disodium Phenyl Phosphate Hydrate 98.0+%, TCI America™

Catalog No. p-7134488
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
P020825G 25 g
P0208500G 500 g
2 options

Catalog No. P020825G

Supplier: TCI America P020825G

Only null left

Chemical Identifiers

CAS 3279-54-7
Molecular Formula C6H5Na2O4P
Molecular Weight (g/mol) 218.06
MDL Number MFCD00002133
InChI Key TYJOJLOWRIQYQM-UHFFFAOYSA-L
Synonym disodium phenyl phosphate, phenyl phosphate disodium salt, disodium phenylphosphate, phenyldisodium phosphate, phosphoric acid, monophenyl ester, disodium salt, phenyl disodium orthophosphate, phenyl phosphate, phosphoric acid, monophenyl ester, sodium salt 1:2, disodium phenyl phosphate dihydrate, phenyl phosphate disodium salt dihydrate
PubChem CID 76771
IUPAC Name disodium phenyl phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 218.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Phenyl Phosphate Hydrate
EINECSNumber (9)-1190
TSCA Yes