missing translation for 'onlineSavingsMsg'
Learn More

Disodium Phenyl Phosphate Hydrate 98.0+%, TCI America™

Catalog No. P020825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P020825G 25 g
P0208500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P020825G Supplier TCI America Supplier No. P020825G

Chemical Identifiers

CAS 3279-54-7
Molecular Formula C6H5Na2O4P
Molecular Weight (g/mol) 218.06
MDL Number MFCD00002133
InChI Key TYJOJLOWRIQYQM-UHFFFAOYSA-L
Synonym disodium phenyl phosphate, phenyl phosphate disodium salt, disodium phenylphosphate, phenyldisodium phosphate, phosphoric acid, monophenyl ester, disodium salt, phenyl disodium orthophosphate, phenyl phosphate, phosphoric acid, monophenyl ester, sodium salt 1:2, disodium phenyl phosphate dihydrate, phenyl phosphate disodium salt dihydrate
PubChem CID 76771
IUPAC Name disodium phenyl phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 218.06
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium Phenyl Phosphate Hydrate
EINECSNumber (9)-1190
TSCA Yes