missing translation for 'onlineSavingsMsg'
Learn More

Disodium Guanylate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014598
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014598 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014598 Supplier Merck Emd Millipore Supplier No. PHR15141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Disodium Guanylate is a flavor enhancer, obtained via chemical synthesis. It is used in combination with monosodium glutamate and disodium inosinate and helps in contributing to a fundamental taste sensation called savory in both animal and vegetable foods. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 5550-12-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C10H12N5 Na2O8 P · xH2 O
MDL Number MFCD00151238
Synonym Guanosine 5′-monophosphate disodium salt hydrate; 5′-GMP-Na2; 5′-Guanylic acid disodium salt hydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 407.18 (anhydrous basis)
Grade Certified Reference Material