missing translation for 'onlineSavingsMsg'
Learn More

Disodium DL-Malate Hydrate 98.0+%, TCI America™

Catalog No. p-7131935
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M002625G 25 g
M0026500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M002625G Supplier TCI America Supplier No. M002625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 676-46-0
Molecular Formula C4H4Na2O5
Molecular Weight (g/mol) 178.051
MDL Number MFCD00012466
InChI Key WPUMTJGUQUYPIV-UHFFFAOYSA-L
Synonym disodium malate, sodium malate, disodium dl-malate, dl-malic acid disodium salt, disodium 2-hydroxybutanedioate, natriummalat german, sodium dl-malate, butanedioic acid, hydroxy-, disodium salt, malic acid, disodium salt, sodium dl-maleate
PubChem CID 8736
IUPAC Name disodium;2-hydroxybutanedioate
SMILES C(C(C(=O)[O-])O)C(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 178.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium DL-Malate Hydrate
EINECSNumber (2)-1449
TSCA No