missing translation for 'onlineSavingsMsg'
Learn More

Disodium beta-Glycerophosphate Pentahydrate 95.0+%, TCI America™

Supplier:  TCI America G0097500G

 View more versions of this product

Catalog No. G0097500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Disodium beta-Glycerophosphate Pentahydrate
102°C
C3H19Na2O11P
500 g
VWVLGHQKEJESPW-UHFFFAOYSA-N
[(1,3-dihydroxypropan-2-yl)oxy]phosphonic acid pentahydrate disodium
131846916
≥95.0% (T)
13408-09-8
White
MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135
unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate
O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O
308.13
216.04
Crystalline Powder

Chemical Identifiers

13408-09-8
308.13
VWVLGHQKEJESPW-UHFFFAOYSA-N
131846916
O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O
C3H19Na2O11P
MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135
unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate
[(1,3-dihydroxypropan-2-yl)oxy]phosphonic acid pentahydrate disodium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1905&(2)-1990

RTECSNumber : UA0600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions