missing translation for 'onlineSavingsMsg'
Learn More

Disodium beta-Glycerophosphate Pentahydrate 95.0+%, TCI America™

Catalog No. G009725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G009725G 25 g
G0097500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G009725G Supplier TCI America Supplier No. G009725G

Chemical Identifiers

CAS 13408-09-8
Molecular Formula C3H19Na2O11P
Molecular Weight (g/mol) 308.13
MDL Number MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135
InChI Key VWVLGHQKEJESPW-UHFFFAOYSA-N
Synonym unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate
PubChem CID 131846916
IUPAC Name [(1,3-dihydroxypropan-2-yl)oxy]phosphonic acid pentahydrate disodium
SMILES O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O

Specifications

Melting Point 102°C
Color White
Quantity 25 g
Formula Weight 216.04
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium beta-Glycerophosphate Pentahydrate
EINECSNumber (2)-1905&(2)-1990
RTECSNumber UA0600000
TSCA Yes