missing translation for 'onlineSavingsMsg'
Learn More

Disodium beta-Glycerophosphate Pentahydrate 95.0+%, TCI America™

Catalog No. D39641G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39641G 1 g
D39645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39641G Supplier TCI America Supplier No. D39641G

Chemical Identifiers

CAS 13408-09-8
Molecular Formula C3H19Na2O11P
Molecular Weight (g/mol) 308.13
MDL Number MFCD00149083,MFCD00002135,MFCD00149083,MFCD00149084,MFCD00002135
InChI Key VWVLGHQKEJESPW-UHFFFAOYSA-N
Synonym unii-5jiy5mmx0e, 5jiy5mmx0e, sodium 2-glycerophosphate pentahydrate, sodium beta-glycerophosphate pentahydrate, sodium 1,3-dihydroxypropan-2-yl phosphate pentahydrate, glycerol 2-phosphate, c3h7na2o6p.5h2o, beta-glycerophosphate, disodium salt, pentahydrate, beta-glycerol phosphate disodium salt pentahydrate, beta-glycerophosphoric acid disodium salt pentahydrate
PubChem CID 131846916
IUPAC Name [(1,3-dihydroxypropan-2-yl)oxy]phosphonic acid pentahydrate disodium
SMILES O.O.O.O.O.[Na].[Na].OCC(CO)OP(O)(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 216.04
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium beta-Glycerophosphate Pentahydrate [for Biochemical Research]
EINECSNumber (2)-1990&(2)-1905
RTECSNumber UA0600000
TSCA Yes
Recommended Storage Refrigerator