missing translation for 'onlineSavingsMsg'
Learn More

Disodium Bathocuproinedisulfonate [for Determination of Cu in Blood], TCI America™

Catalog No. B0985100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0985100MG 100 mg
B09851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0985100MG Supplier TCI America Supplier No. B0985100MG

Chemical Identifiers

CAS 52698-84-7
Molecular Formula C26H18N2Na2O6S2
Molecular Weight (g/mol) 564.54
MDL Number MFCD00149974
InChI Key RNGKZLRAVYPLJC-UHFFFAOYSA-L
Synonym disodium bathocuproinedisulfonate, sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium bathocuproinedisulfonate, bathocuproinedisulfonic acid sodium salt, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate disodium salt, dipotassium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
PubChem CID 15678335
IUPAC Name disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
SMILES [Na+].[Na+].CC1=NC2=C(C=CC3=C2N=C(C)C(=C3C2=CC=CC=C2)S([O-])(=O)=O)C(C2=CC=CC=C2)=C1S([O-])(=O)=O

Specifications

Quantity 100 mg
Formula Weight 564.54
Physical Form Crystalline Powder
Chemical Name or Material Disodium Bathocuproinedisulfonate [for Determination of Cu in Blood]