missing translation for 'onlineSavingsMsg'
Learn More

Disodium 4-Nitrophenyl Phosphate Hexahydrate 98.0+%, TCI America™

Catalog No. n02411g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
25 g
Catalog No. Quantity
N02411G 1 g
N024125G 25 g
2 options

Catalog No. N02411G

Supplier: TCI America N02411G

Only null left

Chemical Identifiers

CAS 333338-18-4
Molecular Formula C6H16NNa2O12P
Molecular Weight (g/mol) 371.142
MDL Number MFCD00066288
InChI Key KAKKHKRHCKCAGH-UHFFFAOYSA-L
Synonym Phosphoric Acid 4-Nitrophenyl Ester Disodium Salt
PubChem CID 2723710
IUPAC Name disodium;(4-nitrophenyl) phosphate;hexahydrate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OP(=O)([O-])[O-].O.O.O.O.O.O.[Na+].[Na+]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 263.05
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4-Nitrophenyl Phosphate Hexahydrate
EINECSNumber (9)-0941
TSCA Yes
Recommended Storage Refrigerator