missing translation for 'onlineSavingsMsg'
Learn More

Disodium 4-Formylbenzene-1,3-disulfonate 98.0+%, TCI America™

Catalog No. b000825g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
B000825G 25 g
1 options

Catalog No. B000825G

Supplier: TCI America B000825G

Only null left

Specifications

CAS 33513-44-9
Color White
Molecular Formula C7H4Na2O7S2
Quantity 25 g
MDL Number MFCD00043913
Synonym sodium 4-formylbenzene-1,3-disulfonate, benzaldehyde-2,4-disulfonic acid disodium salt, disodium 4-formylbenzene-1,3-disulfonate, disodium 4-formylbenzene-1,3-disulphonate, 1,3-benzenedisulfonic acid, 4-formyl-, disodium salt, 1,3-benzenedisulfonic acid, 4-formyl-, sodium salt 1:2, sodium 4-formylbenzene-1,3-disulfonate hydrate, 4-formyl-1,3-benzenedisulfonic acid, disodium salt, acmc-209i1i, benzaldehyde 2,4 disulfonic acid
InChI Key UUKHCUPMVISNFW-UHFFFAOYSA-L
SMILES C1=CC(=C(C=C1S(=O)(=O)[O-])S(=O)(=O)[O-])C=O.[Na+].[Na+]
IUPAC Name disodium;4-formylbenzene-1,3-disulfonate
Molecular Weight (g/mol) 310.202
PubChem CID 169512
Formula Weight 310.20
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4-Formylbenzene-1,3-disulfonate
Show More Show Less
EINECSNumber (3)-2108
TSCA Yes