missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium 4,4'-Bis(2-sulfonatostyryl)biphenyl 97.0+%, TCI America™
$141.67 - $367.22
Chemical Identifiers
| CAS | 27344-41-8 |
|---|---|
| Molecular Formula | C28H20Na2O6S2 |
| Molecular Weight (g/mol) | 562.56 |
| MDL Number | MFCD00318874,MFCD00318874 |
| InChI Key | PMPJQLCPEQFEJW-GNTLFSRWSA-L |
| PubChem CID | 87075690 |
| IUPAC Name | disodium 2-[(1Z)-2-{4'-[(1Z)-2-(2-sulfonatophenyl)ethenyl]-[1,1'-biphenyl]-4-yl}ethenyl]benzene-1-sulfonate |
| SMILES | [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1\C=C/C1=CC=C(C=C1)C1=CC=C(\C=C/C2=CC=CC=C2S([O-])(=O)=O)C=C1 |
Chemical Identifiers
| 27344-41-8 | |
| 562.56 | |
| PMPJQLCPEQFEJW-GNTLFSRWSA-L | |
| disodium 2-[(1Z)-2-{4'-[(1Z)-2-(2-sulfonatophenyl)ethenyl]-[1,1'-biphenyl]-4-yl}ethenyl]benzene-1-sulfonate |
| C28H20Na2O6S2 | |
| MFCD00318874,MFCD00318874 | |
| 87075690 | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1\C=C/C1=CC=C(C=C1)C1=CC=C(\C=C/C2=CC=CC=C2S([O-])(=O)=O)C=C1 |
Specifications
| 27344-41-8 | |
| C28H20Na2O6S2 | |
| 25 g | |
| PMPJQLCPEQFEJW-GNTLFSRWSA-L | |
| disodium 2-[(1Z)-2-{4'-[(1Z)-2-(2-sulfonatophenyl)ethenyl]-[1,1'-biphenyl]-4-yl}ethenyl]benzene-1-sulfonate | |
| 87075690 | |
| ≥97.0% (HPLC) | |
| Disodium 4,4′-Bis(2-sulfonatostyryl)biphenyl |
| Yellow | |
| MFCD00318874,MFCD00318874 | |
| 4,4′C-Bis(2-sulfonatostyryl)biphenyl Disodium Salt | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1\C=C/C1=CC=C(C=C1)C1=CC=C(\C=C/C2=CC=CC=C2S([O-])(=O)=O)C=C1 | |
| 562.56 | |
| 562.56 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-4880
TSCA : Yes