missing translation for 'onlineSavingsMsg'
Learn More

Disodium 4,4'-Bis(2-sulfonatostyryl)biphenyl 97.0+%, TCI America™

Catalog No. D325125G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D325125G 25 g
D3251100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D325125G Supplier TCI America Supplier No. D325125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27344-41-8
Molecular Formula C28H20Na2O6S2
Molecular Weight (g/mol) 562.56
MDL Number MFCD00318874,MFCD00318874
InChI Key PMPJQLCPEQFEJW-GNTLFSRWSA-L
PubChem CID 87075690
IUPAC Name disodium 2-[(1Z)-2-{4'-[(1Z)-2-(2-sulfonatophenyl)ethenyl]-[1,1'-biphenyl]-4-yl}ethenyl]benzene-1-sulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1\C=C/C1=CC=C(C=C1)C1=CC=C(\C=C/C2=CC=CC=C2S([O-])(=O)=O)C=C1

Specifications

Color Yellow
Quantity 25 g
Synonym 4,4′C-Bis(2-sulfonatostyryl)biphenyl Disodium Salt
Formula Weight 562.56
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 4,4′-Bis(2-sulfonatostyryl)biphenyl
EINECSNumber (5)-4880
TSCA Yes