missing translation for 'onlineSavingsMsg'
Learn More

Disodium 1,3-Propanedisulfonate 97.0+%, TCI America™

Catalog No. P12055G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P12055G Supplier TCI America Supplier No. P12055G

Chemical Identifiers

CAS 36589-58-9
Molecular Formula C3H6Na2O6S2
Molecular Weight (g/mol) 248.175
MDL Number MFCD00191483
InChI Key DKGJFKPIUSHDIT-UHFFFAOYSA-L
Synonym eprodisate disodium, disodium 1,3-propanedisulfonate, kiacta, disodium propane-1,3-disulfonate, eprodisate sodium, unii-5x0d9h16iu, 1,3-propanedisulfonic acid disodium salt, eprodisate disodium usan, 1,3-propanedisulfonic acid, disodium salt
PubChem CID 6451143
IUPAC Name disodium;propane-1,3-disulfonate
SMILES C(CS(=O)(=O)[O-])CS(=O)(=O)[O-].[Na+].[Na+]

Specifications

Color White
Quantity 5 g
Formula Weight 248.17
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 1,3-Propanedisulfonate