missing translation for 'onlineSavingsMsg'
Learn More

Disodium 1,3-Benzenedisulfonate 55.0+%, TCI America™

Catalog No. B002325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B002325G 25 g
B0023500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B002325G Supplier TCI America Supplier No. B002325G

Chemical Identifiers

CAS 831-59-4
Molecular Formula C6H4Na2O6S2
Molecular Weight (g/mol) 282.19
MDL Number MFCD00007492
InChI Key XWPWZOJBTOJEGW-UHFFFAOYSA-L
Synonym disodium 1,3-benzenedisulfonate, disodium benzene-1,3-disulfonate, 1,3-benzenedisulfonic acid, disodium salt, benzene-1,3-disulfonic acid disodium salt, 1,3-benzenedisulfonic acid disodium salt, disodium benzene-1,3-disulphonate, disodium m-benzenedisulfonate, sodium benzene-1,3-disulfonate, 1,3-benzenedisulfonic acid, sodium salt 1:2, m-benzenedisulfonic acid, disodium salt
PubChem CID 71577
IUPAC Name disodium benzene-1,3-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1)S([O-])(=O)=O

Specifications

Quantity 25 g
Formula Weight 282.19
Percent Purity ≥55.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Disodium 1,3-Benzenedisulfonate
EINECSNumber (3)-1888
TSCA Yes