missing translation for 'onlineSavingsMsg'
Learn More

Dentec Safety™ Comfort-Air™ Series 300 Diskit™ P100 Complete Respirators

Catalog No. 19222448
Change view
Click to view available options
Size:
Medium/Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19222448 Medium/Large
19222447 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19222448 Supplier Dentec Safety™ Supplier No. DTC6516575LDISKP100ZAM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Series 300 thermoplastic half-mask respirators with Diskit™ P100 filter assembly are ideal for welding, dust and oil mist applications.

  • Series 300 thermoplastic respirators complete with Diskit™ P100 filter assembly
  • Extended field of vision
  • Large inhalation valves allows users to inhale a larger volume of air per breath, reducing breathing resistance
  • Exhalation valve is strategically positioned under the nostrils and in front of the mouth to ensure hot, exhaled air exits the mask immediately
  • Exhalation valve diaphragm has channels that act like a parachute to traps exhaled air and lift the diaphragm off the valve seat with very little effort, creating more space for air to exit the mask, thereby reducing heat build up inside the mask
  • Rolled sealing flange provides comfort, improves fit, and does not require user to adjust straps excessively
  • Textured sealing area reduces risk of the mask slipping on the face
  • Enhanced harness assembly features:
    • Metal tabs at end to prevent nylon strap fraying and allows for adjustment when wearing gloves
    • Wide nylon strap is more comfortable against the skin and maintains elasticity
    • 4-point swiveling adjustments allows the mask to be adjusted
    • Cradle suspension fits comfortably under suspension systems for hard hats, grinding and welding helmets
  • 3- position adjustment allows users to adjust cartridge to three different positions

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Gray
Filter Type P100
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Medium/Large
With Exhalation Valve Yes
Facepiece Material Thermoplastic Elastomer
For Use With (Equipment) Cartridges/Diskits
Model 3-ML-DISKP100ZAM
Packaging Type Display Bag
Length (Metric) 10.16 cm
Width (Metric) 16.51 cm
Height (Metric) 12.7 cm
Show More Show Less