Learn More
Berkshire™ EZ Bleach™ Disinfectant Tablets
Rapidly dissolving 48.21% sodium dichloro-s-triazinetrione tablets.
Supplier:  Berkshire™ EZBDT101CA
Catalog No. 19266709
Only null left
Description
Description
  • NaDCC
  • Each rapidly dissolving 48.21% sodium dichloro-s-triazinetrione tablet is the equivalent of 29% available chlorine, a broad spectrum and sporicidal disinfectant
  • 1 tub of 900 tablets will produce 190 gallons of liquid bleach
Specifications
Specifications
Non-sterile
900 Tablets/Tub
Product Suggestions
Product Suggestions