missing translation for 'onlineSavingsMsg'
Learn More

Discovery Education™ If We Had No Moon

Catalog No. S95480DVD
Change view
Click to view available options
Grade:
3 to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S95480DVD 3 to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S95480DVD Supplier Discovery Education™ Supplier No. 904110

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Experts reveal the enormous impact the moon has on Earth

Chances are there would be no life on Earth if there were no moon. Scientists simulate how an ancient rogue planet collided with Earth millions of years ago and created our moon, which impacts our weather, gravitational pull, rotation, and living creatures.

  • Grades 9-12
  • 24 mins.

Specifications

Grade 3 to 5
Copyright Year 2007
Includes Marc Record
Name If We Had No Moon DVD
Run Time 56 min.
Format DVD