missing translation for 'onlineSavingsMsg'
Learn More

Discovery Education™ Earth Science: The Basics, Grades 3 to 5

Catalog No. S90447DVD
Change view
Click to view available options
Grade:
3 to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S90447DVD 3 to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90447DVD Supplier Discovery Education™ Supplier No. 880518

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

From oceans and volcanoes to fossils and storms

Introduce young students to a wide variety of Earth science topics with these segments drawn from this award-winning series.

The title includes six scientific literacy segments to aid classroom teachers along with the following 12 student video segments:

  • Introducing Earth
  • When Earth Erupts
  • Earthquake Zone
  • Erosion
  • The Rock Cycle
  • Granite Rocks
  • Precious Minerals
  • Changes in Weather and Climate
  • Violent Weather
  • Making Sea Water Fresh
  • Earth's Atmosphere
  • Ocean Motions

Order Info

Grade 3 to 5. Running time: 3 hours.

Specifications

Grade 3 to 5
Run Time 180 min.
Copyright Year 2005
Name Earth Science: The Basics Grades 3-5 DVD
Includes Marc Record, Teacher's Guide
Format DVD