missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Discovery™ DSC-Si SPE Tube, Bed Wt. 1 g

Catalog No. 111020815
Change view
Click to view available options
Bed Weight:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight
111020815 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020815 Supplier MilliporeSigma™ Supelco™ Supplier No. 52656U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

Retention Mechanism: Normal phaseSample Matrix Compatibility: Organic solvents, oils, and lipids. Unbonded acid washed silica sorbent ideal for normal phase SPE and other modified flash techniques. Often used to separate or remove structurally similar molecules through successive elutions with increasingly polar solutions. The most polar normal phase sorbent available. Excellent capacity for purifying solution phase combinatorial chemistry reactions when removing target molecules from reaction by-products and excess reagents

Specifications

Column Phase DSC-Si
Column Type SPE
Bed Weight 1 g
For Use With (Application) Food and Beverages
Product Line Discovery
Type Solid Phase Extraction Column
Volume (Metric) 6 mL
Material PE Frit
Pore Size 70 Å
Quantity 30/Pk.
Surface Area (Metric) 480 m2/g
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC