missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Discovery™ DSC-SCX SPE Tube, Bed Wt. 2 g

Catalog No. 111017084
Change view
Click to view available options
Bed Weight:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight
111017084 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017084 Supplier MilliporeSigma™ Supelco™ Supplier No. 52690U

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

Retention Mechanism: Cation exchangeSample Matrix Compatibility: Organic or aqueous solutions. A polymerically bonded, benzene sulfonic acid functional group, pKa (< 1.0). Counter ion is H+. Silica support allows for use with very organic solvents (no shrinking/swelling). Excellent capacity (0.8 meq/g) for cleaning up solution phase combinatorial chemistry reactions (removing target molecules from reaction by-products and excess reagents). The presence of the benzene ring offers some mixed-mode capabilities (hydrophobic interactions) that should be considered when extracting cations from aqueous matrices

Specifications

Column Capacity (Metric) 0.8 meq/g total capacity
Column Phase DSC-SCX
Column Type SPE
Bed Weight 2 g
For Use With (Application) Food and Beverages
Product Line Discovery
Type Solid Phase Extraction Column
Volume (Metric) 12 mL
Material Polypropylene
Pore Size 70 Å
Quantity 20/Pk.
Surface Area (Metric) 480 m2/g
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC