missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Discovery™ DSC-8 SPE Tube, Bed Wt. 50 mg

Catalog No. 111009754
Change view
Click to view available options
Bed Weight:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight
111009754 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111009754 Supplier MilliporeSigma™ Supelco™ Supplier No. 52703U

Solid phase extraction (SPE) is a technique commonly used in laboratories to isolate analytes of interest from complex matrices.

Retention Mechanism: Reversed-phaseSample Matrix Compatibility: Aqueous solutions (biological fluids, water). Monomerically bonded, octyl (9 %C), endcapped; lower carbon content than DSC-18Lt. Used to elute very large hydrophobic molecules too strongly retained on DSC-18 or DSC-18Lt. Use this less retentive phase for the rapid release of hydrophobic molecules using weaker organic solvents at lower volumes. Inorganic buffers of sufficient ionic strength may be used for elution

Specifications

Column Phase DSC-8
Column Type SPE
Bed Weight 50 mg
For Use With (Application) Food and Beverages
Product Line Discovery
Type Solid Phase Extraction Column
Volume (Metric) 1 mL
Material Polypropylene
Pore Size 70 Å
Quantity 108/Pk.
Surface Area (Metric) 480 m2/g
Show More Show Less

Discovery is a registered trademark of Sigma-Aldrich Co. LLC