missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Discharge Tubes for Dispensette™ S Bottletop Dispensers with standard valves

Catalog No. 14380898
Change view
Click to view available options
For Use With (Equipment):
Dispensette S Organic, nominal volumes 25, 50 and 10 mL
Dispensette S Organic, nominal volumes 25, 50 and 100 mL
Dispensette S Organic, nominal volumes 5 and 10 mL
Dispensette S Trace Analysis, Platinum-Iridium
Dispensette S Trace Analysis, Tantalum
Dispensette S, 1, 2, 5 and 10 mL nominal volumes
Dispensette S, 25, 50 and 100 mL nominal volumes
Dispensette S, nominal volumes 25, 50 and 100 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
14380898 Dispensette S Trace Analysis, Tantalum
14380890 Dispensette S, 1, 2, 5 and 10 mL nominal volumes
14380892 Dispensette S, 25, 50 and 100 mL nominal volumes
14380893 Dispensette S, nominal volumes 25, 50 and 100 mL
14380894 Dispensette S Organic, nominal volumes 5 and 10 mL
14380895 Dispensette S Organic, nominal volumes 25, 50 and 100 mL
14380896 Dispensette S Organic, nominal volumes 25, 50 and 10 mL
14380897 Dispensette S Trace Analysis, Platinum-Iridium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14380898 Supplier BRANDTECH™ BRAND™ Supplier No. 708024

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Discharge tubes to fit Dispensette™ made 2016 and later

  • Discharge tubes come in two sizes: For instruments with nominal capacity 1 mL, 2 mL, 5 mL, and 10 mL; and for instruments with nominal capacity of 25 mL, 50 mL and 100 mL
  • Choose from standard and fine tip models
  • Dispensette™ S dispensers have red trim
  • Dispensette™ S Organic dispensers have yellow trim

Specifications

Product Type Discharge Tube
Autoclavable Not Autoclavable
Color Translucent/White
Material ETFE, PTFE, Polypropylene
For Use With (Application) Dispensing liquid
For Use With (Equipment) Dispensette S Trace Analysis, Tantalum
Includes Discharge Tube, Safety Cap