Perkin Elmer Canada Infrared Window for Microscopic FTIR Analysis - NaCl, 2 mm
Catalog No. 509045892
Infrared windows allow for support of microsamples for infrared analysis with microscopes, such as PerkinElmer's Spotlight Systems. These windows are available in the following infrared crystals: BaF2, KBr, NaCl, and ZnSe.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Infrared windows allow for support of microsamples for infrared analysis with microscopes, such as PerkinElmer's Spotlight Systems. These windows are available in the following infrared crystals: BaF2, KBr, NaCl, and ZnSe.