missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Disarticulated Human Rib Bones
Anatomically correct disarticulated human rib bones are replicas but provide an incredible amount of realism and detail for examining the human skeleton
$47.10 - $72.13
Specifications
| Color | Natural |
|---|---|
| For Use With (Application) | Examining The Human Skeleton |
| Material | PVC |
| Product Type | Anatomical Model, Human |
Description
- Anatomically correct bones
- Realistic details, textures, and bony landmarks
- Ideal for educational and medical demonstrations
- Use for individual study or as replacement for missing/damaged pieces to a full skeletal model set
- Made of durable and sturdy PVC plastic
- Natural size and color
- Bones are marked for easy demonstration
- Washable
Specifications
| Natural | |
| Examining The Human Skeleton | |
| PVC | |
| Anatomical Model, Human |