missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Disarticulated Human Rib Bones

Catalog No. S27455
Change view
Click to view available options
Includes:
12 Bones (Half Set)
24 Bones (Full Set)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S27455 12 Bones (Half Set)
S27454 24 Bones (Full Set)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27455 Supplier Eisco™ Supplier No. AMCH1043AS

Anatomically correct disarticulated human rib bones are replicas but provide an incredible amount of realism and detail for examining the human skeleton

  • Anatomically correct bones
  • Realistic details, textures, and bony landmarks
  • Ideal for educational and medical demonstrations
  • Use for individual study or as replacement for missing/damaged pieces to a full skeletal model set
  • Made of durable and sturdy PVC plastic
  • Natural size and color
  • Bones are marked for easy demonstration
  • Washable

Specifications

Color Natural
For Use With (Application) Examining The Human Skeleton
Includes 12 Bones (Half Set)
Material PVC
Product Type Anatomical Model, Human